siRNA Details
| Gene Symbol: Tbcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108200.2 | NP_001101670.1 | 1 | 939 | ||
| GenBank mRNA | BC166467.1 | - | 1 | 939 | 9035211 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbcc| Gene Symbol: Tbcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108200.2 | NP_001101670.1 | 1 | 939 | ||
| GenBank mRNA | BC166467.1 | - | 1 | 939 | 9035211 | |