siRNA Details
Gene Symbol: Mad2l1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009699.1 | NP_001009699.1 | 3 | 438 | ||
GenBank mRNA | BC088268.1 | - | 3 | 438 | 7379293 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mad2l1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009699.1 | NP_001009699.1 | 3 | 438 | ||
GenBank mRNA | BC088268.1 | - | 3 | 438 | 7379293 |