siRNA Details
| Gene Symbol: Cd2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181475.2 | NP_852140.1 | 1 | 152 | ||
| GenBank mRNA | AY114147.1 | - | 1 | 148 | ||
| AY205155.1 | - | 1 | 152 | |||
| CB545525.1 | - | 272 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd2ap| Gene Symbol: Cd2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181475.2 | NP_852140.1 | 1 | 152 | ||
| GenBank mRNA | AY114147.1 | - | 1 | 148 | ||
| AY205155.1 | - | 1 | 152 | |||
| CB545525.1 | - | 272 | ||||