siRNA Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033705.1 | NP_001028877.1 | 10 | 2361 | ||
| GenBank mRNA | BC101930.1 | - | 10 | 2361 | 7938755 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp451| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033705.1 | NP_001028877.1 | 10 | 2361 | ||
| GenBank mRNA | BC101930.1 | - | 10 | 2361 | 7938755 | |