siRNA Details
| Gene Symbol: Neurl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014100.1 | NP_001014122.1 | 4 | 1344 | ||
| XM_008767025.1 | XP_008765247.1 | 5 | 1640 | |||
| XM_008767026.1 | XP_008765248.1 | 5 | 1418 | |||
| GenBank mRNA | BC088198.1 | - | 4 | 1344 | 7376146 | |

Neurl3
Genomic Map