siRNA Details
| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244855.2 | XP_006244917.1 | 38 | 5045 | ||
| XM_006244856.2 | XP_006244918.1 | 38 | 5126 | |||
| XM_006244858.2 | XP_006244920.1 | 37 | 6963 | |||
| XM_237056.8 | XP_237056.4 | 38 | 5045 | |||
| GenBank mRNA | FQ223997.1 | - | 10 | 1820 | ||
| FQ224326.1 | - | 10 | 1820 | |||

Tmem131
Genomic Map