siRNA Details
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108213.1 | NP_001101683.1 | 5 | 1134 | ||
XM_006244819.2 | XP_006244881.1 | 6 | 1866 | |||
XM_006244820.2 | XP_006244882.1 | 6 | 2486 | |||
XM_006244821.2 | XP_006244883.1 | 5 | 1668 | |||
XM_008767030.1 | XP_008765252.1 | 2 | 673 |
