siRNA Details
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108213.1 | NP_001101683.1 | 4 | 403 | ||
XM_006244819.2 | XP_006244881.1 | 5 | 1135 | |||
XM_006244820.2 | XP_006244882.1 | 5 | 1755 | |||
XM_006244821.2 | XP_006244883.1 | 4 | 937 |
