siRNA Details
| Gene Symbol: Nck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108216.1 | NP_001101686.1 | 3 | 431 | ||
| XM_006244825.2 | XP_006244887.1 | 3 | 396 | |||
| XM_008767035.1 | XP_008765257.1 | 3 | 2840 | |||
| GenBank mRNA | AY724492.1 | - | 3 | 431 | ||

Nck2
Genomic Map