siRNA Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277228.1 | NP_001264157.1 | 4 | 924 | ||
| GenBank mRNA | BC085943.1 | - | 4 | 924 | ||
| BC168987.1 | - | 4 | 886 | 9030170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelc1| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277228.1 | NP_001264157.1 | 4 | 924 | ||
| GenBank mRNA | BC085943.1 | - | 4 | 924 | ||
| BC168987.1 | - | 4 | 886 | 9030170 | ||