siRNA Details
| Gene Symbol: Ccdc150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191806.1 | NP_001178735.1 | 23 | 2786 | ||
| XM_006244924.2 | XP_006244986.1 | 23 | 2942 | |||
| XM_008767094.1 | XP_008765316.1 | 24 | 3008 | |||
| XM_008767095.1 | XP_008765317.1 | 23 | 2942 | |||
| XM_008767096.1 | XP_008765318.1 | 23 | 2942 | |||
| XM_008767097.1 | XP_008765319.1 | 23 | 2942 | |||

Ccdc150
Genomic Map