siRNA Details
| Gene Symbol: Mars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750701.3 | XP_003750749.1 | 1 | 1190 | ||
| Gene Symbol: LOC100911305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750699.3 | XP_003750747.1 | 1 | 1190 | ||

Mars2
Genomic Map