siRNA Details
| Gene Symbol: Als2cr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014101.1 | NP_001014123.1 | 9 | 1011 | ||
| GenBank mRNA | BC079241.1 | - | 9 | 1011 | 7132659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Als2cr12| Gene Symbol: Als2cr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014101.1 | NP_001014123.1 | 9 | 1011 | ||
| GenBank mRNA | BC079241.1 | - | 9 | 1011 | 7132659 | |