siRNA Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025710.1 | NP_001020881.1 | 7, 8 | 632 | ||
| XM_006244983.2 | XP_006245045.1 | 6, 7 | 639 | |||
| XM_006244984.2 | XP_006245046.1 | 6, 7 | 638 | |||
| XM_006244985.2 | XP_006245047.1 | 7, 8 | 672 | |||
| XM_008767100.1 | XP_008765322.1 | 6, 7 | 1276 | |||
| XM_008767101.1 | XP_008765323.1 | 7, 8 | 888 | |||
| XM_008767102.1 | XP_008765324.1 | 7, 8 | 673 | |||
| XM_008767103.1 | XP_008765325.1 | 6, 7 | 1187 | |||
| XM_008767104.1 | XP_008765326.1 | 6, 7 | 636 | |||
| XM_008767105.1 | XP_008765327.1 | 6, 7 | 639 | |||
| GenBank mRNA | BC097998.1 | - | 7, 8 | 632 | 7453086 | |

Fam126b
Genomic Map