siRNA Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025710.1 | NP_001020881.1 | 13 | 1540 | ||
| XM_006244983.2 | XP_006245045.1 | 13 | 1715 | |||
| XM_006244984.2 | XP_006245046.1 | 12 | 1546 | |||
| XM_006244985.2 | XP_006245047.1 | 13 | 1580 | |||
| XM_008767100.1 | XP_008765322.1 | 13 | 2352 | |||
| XM_008767101.1 | XP_008765323.1 | 14 | 1964 | |||
| XM_008767102.1 | XP_008765324.1 | 14 | 1749 | |||
| XM_008767103.1 | XP_008765325.1 | 13 | 2263 | |||
| XM_008767104.1 | XP_008765326.1 | 13 | 1712 | |||
| GenBank mRNA | BC097998.1 | - | 13 | 1540 | 7453086 | |

Fam126b
Genomic Map