siRNA Details
| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008523.1 | NP_001008523.1 | 23 | 2609 | ||
| XM_006244987.2 | XP_006245049.1 | 23 | 2580 | |||
| GenBank mRNA | AY665587.1 | - | 23 | 2609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aox4| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008523.1 | NP_001008523.1 | 23 | 2609 | ||
| XM_006244987.2 | XP_006245049.1 | 23 | 2580 | |||
| GenBank mRNA | AY665587.1 | - | 23 | 2609 | ||