siRNA Details
| Gene Symbol: Spats2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014102.1 | NP_001014124.1 | 7 | 999 | ||
| XM_006244990.2 | XP_006245052.1 | 8 | 871 | |||
| XM_006244991.2 | XP_006245053.1 | 8 | 872 | |||
| XM_006244992.2 | XP_006245054.1 | 9 | 976 | |||
| XM_006244993.2 | XP_006245055.1 | 7 | 664 | |||
| XM_008767107.1 | XP_008765329.1 | 7 | 664 | |||
| XM_008767108.1 | XP_008765330.1 | 8 | 1016 | |||
| XM_008767109.1 | XP_008765331.1 | 8 | 829 | |||
| XM_008767110.1 | XP_008765332.1 | 7 | 809 | |||
| GenBank mRNA | BC085874.1 | - | 7 | 999 | 7191441 | |

Spats2l
Genomic Map