siRNA Details
| Gene Symbol: Spats2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014102.1 | NP_001014124.1 | 2, 3 | 338 | ||
| XM_006244990.2 | XP_006245052.1 | 3, 4 | 210 | |||
| XM_006244991.2 | XP_006245053.1 | 3, 4 | 211 | |||
| XM_006244992.2 | XP_006245054.1 | 4, 5 | 315 | |||
| XM_006244993.2 | XP_006245055.1 | 3, 4 | 210 | |||
| XM_008767107.1 | XP_008765329.1 | 3, 4 | 210 | |||
| XM_008767108.1 | XP_008765330.1 | 3, 4 | 355 | |||
| XM_008767109.1 | XP_008765331.1 | 3, 4 | 168 | |||
| XM_008767110.1 | XP_008765332.1 | 2, 3 | 148 | |||
| GenBank mRNA | BC085874.1 | - | 2, 3 | 338 | 7191441 | |

Spats2l
Genomic Map