siRNA Details
| Gene Symbol: Cyp20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199401.1 | NP_955433.1 | 10 | 1089 | ||
| GenBank mRNA | BC061716.1 | - | 10 | 1089 | 5598443 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp20a1| Gene Symbol: Cyp20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199401.1 | NP_955433.1 | 10 | 1089 | ||
| GenBank mRNA | BC061716.1 | - | 10 | 1089 | 5598443 | |