siRNA Details
| Gene Symbol: Ccnyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767164.1 | XP_008765386.1 | 5, 6 | 474 | ||
| XM_008767165.1 | XP_008765387.1 | 4, 5 | 564 | |||
| XM_008767166.1 | XP_008765388.1 | 5, 6 | 473 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001097644, AK132782.

Ccnyl1
Genomic Map