siRNA Details
| Gene Symbol: Ccnyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767164.1 | XP_008765386.1 | 3 | 319 | ||
| XM_008767165.1 | XP_008765387.1 | 2 | 409 | |||
| XM_008767166.1 | XP_008765388.1 | 3 | 318 | |||
| XM_008767167.1 | XP_008765389.1 | 2 | 414 | |||

Ccnyl1
Genomic Map