siRNA Details
| Gene Symbol: LOC689991 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767349.1 | XP_008765571.1 | 51 | 7517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689991| Gene Symbol: LOC689991 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767349.1 | XP_008765571.1 | 51 | 7517 | ||