siRNA Details
| Gene Symbol: Crispld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134963.1 | NP_001128435.1 | 2 | 1015 | ||
| XM_008763477.1 | XP_008761699.1 | 2 | 689 | |||
| GenBank mRNA | BC169118.1 | - | 2 | 654 | 9029822 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crispld1| Gene Symbol: Crispld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134963.1 | NP_001128435.1 | 2 | 1015 | ||
| XM_008763477.1 | XP_008761699.1 | 2 | 689 | |||
| GenBank mRNA | BC169118.1 | - | 2 | 654 | 9029822 | |