siRNA Details
| Gene Symbol: Tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007713.1 | NP_001007714.1 | 3, 4 | 417 | ||
| XM_008767238.1 | XP_008765460.1 | 3, 4 | 2789 | |||
| XM_008767239.1 | XP_008765461.1 | 4, 5 | 440 | |||
| GenBank mRNA | BC079087.1 | - | 3, 4 | 417 | 7123233 | |

Tmbim1
Genomic Map