siRNA Details
| Gene Symbol: Slc11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031658.1 | NP_001026828.1 | 9 | 968 | ||
| XM_006245219.2 | XP_006245281.1 | 10 | 1046 | |||
| XM_006245220.1 | XP_006245282.1 | 10 | 1094 | |||
| GenBank mRNA | BC100653.1 | - | 9 | 968 | 7932285 | |

Slc11a1
Genomic Map