siRNA Details
| Gene Symbol: Vil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108224.2 | NP_001101694.1 | 9, 10 | 1140 | ||
| GenBank mRNA | BC168981.1 | - | 9, 10 | 1140 | 7462629 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vil1| Gene Symbol: Vil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108224.2 | NP_001101694.1 | 9, 10 | 1140 | ||
| GenBank mRNA | BC168981.1 | - | 9, 10 | 1140 | 7462629 | |