siRNA Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127492.1 | NP_001120964.1 | 9 | 3476 | ||
| XM_006245230.2 | XP_006245292.1 | 8 | 3705 | |||
| XM_006245231.2 | XP_006245293.1 | 8 | 4106 | |||
| XM_006245233.2 | XP_006245295.1 | 7 | 3625 | |||
| XM_006245234.2 | XP_006245296.1 | 8 | 3704 | |||
| XM_008767251.1 | XP_008765473.1 | 7 | 4027 | |||

Sphkap
Genomic Map