siRNA Details
| Gene Symbol: Sp140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012133.1 | NP_001012133.1 | 8, 9 | 1464 | ||
| GenBank mRNA | BC081971.1 | - | 8, 9 | 1464 | 7125855 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp140| Gene Symbol: Sp140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012133.1 | NP_001012133.1 | 8, 9 | 1464 | ||
| GenBank mRNA | BC081971.1 | - | 8, 9 | 1464 | 7125855 | |