siRNA Details
| Gene Symbol: B3gnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012134.1 | NP_001012134.1 | 690 | |||
| GenBank mRNA | BC081994.1 | - | 690 | 7130315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gnt7| Gene Symbol: B3gnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012134.1 | NP_001012134.1 | 690 | |||
| GenBank mRNA | BC081994.1 | - | 690 | 7130315 | ||