siRNA Details
| Gene Symbol: Usp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134885.1 | NP_001128357.1 | 3 | 365 | ||
| XM_008767258.1 | XP_008765480.1 | 3 | 931 | |||
| XM_008767259.1 | XP_008765481.1 | 3 | 826 | |||
| XM_008767260.1 | XP_008765482.1 | 3 | 821 | |||
| XM_008767261.1 | XP_008765483.1 | 2 | 1287 | |||
| Gene Symbol: LOC100911860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767354.1 | XP_008765576.1 | 2 | 1287 | ||

Usp40
Genomic Map