siRNA Details
| Gene Symbol: Asb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108232.2 | NP_001101702.1 | 3 | 1004 | ||
| XM_006245409.2 | XP_006245471.1 | 3 | 514 | |||
| XM_006245410.2 | XP_006245472.1 | 3 | 519 | |||
| GenBank mRNA | BC168978.1 | - | 3 | 1004 | 7455775 | |

Asb1
Genomic Map