siRNA Details
| Gene Symbol: Olr1346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000520.1 | NP_001000520.1 | 1 | 280 | ||
| GenBank mRNA | AF091578.1 | - | 1 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1346| Gene Symbol: Olr1346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000520.1 | NP_001000520.1 | 1 | 280 | ||
| GenBank mRNA | AF091578.1 | - | 1 | 70 | ||