siRNA Details
| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025711.1 | NP_001020882.1 | 2, 3 | 321 | ||
| XM_006245533.2 | XP_006245595.1 | 8, 9 | 861 | |||
| XM_006245534.2 | XP_006245596.1 | 7, 8 | 764 | |||
| GenBank mRNA | BC098833.1 | - | 2, 3 | 321 | 7378703 | |

Atg4b
Genomic Map