siRNA Details
| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025711.1 | NP_001020882.1 | 6, 7 | 698 | ||
| XM_006245533.2 | XP_006245595.1 | 12, 13 | 1238 | |||
| XM_006245534.2 | XP_006245596.1 | 11, 12 | 1141 | |||
| GenBank mRNA | BC098833.1 | - | 6, 7 | 698 | 7378703 | |

Atg4b
Genomic Map