siRNA Details
| Gene Symbol: Neu4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108234.1 | NP_001101704.1 | 1494 | |||
| XM_008767376.1 | XP_008765598.1 | 2854 | ||||
| Gene Symbol: NEWGENE_1308624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245535.2 | XP_006245597.1 | 3 | 2854 | ||
| XM_008767394.1 | XP_008765616.1 | 4 | 1475 | |||

Neu4
Genomic Map