siRNA Details
| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108235.1 | NP_001101705.1 | 6, 7 | 1029 | ||
| GenBank mRNA | BC168863.1 | - | 1237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl17| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108235.1 | NP_001101705.1 | 6, 7 | 1029 | ||
| GenBank mRNA | BC168863.1 | - | 1237 | |||