siRNA Details
| Gene Symbol: Fam114a1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_237514.9 | XP_237514.6 | 1 | 1140 | ||
| Gene Symbol: Fam114a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250991.2 | XP_006251053.1 | 9 | 1203 | ||
| XM_008770176.1 | XP_008768398.1 | 9 | 1491 | |||
| XM_008770177.1 | XP_008768399.1 | 9 | 1209 | |||

Fam114a1l1
Genomic Map