siRNA Details
| Gene Symbol: Lpin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108236.1 | NP_001101706.1 | 7 | 1019 | ||
| XM_006245662.2 | XP_006245724.1 | 7 | 3323 | |||
| XM_006245663.2 | XP_006245725.1 | 7 | 4340 | |||
| XM_006245664.2 | XP_006245726.1 | 7 | 1568 | |||
| XM_006245665.2 | XP_006245727.1 | 7 | 3325 | |||
| XM_006245666.2 | XP_006245728.1 | 7 | 1057 | |||

Lpin2
Genomic Map