siRNA Details
| Gene Symbol: Lpin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108236.1 | NP_001101706.1 | 18, 19 | 2534 | ||
| XM_006245662.2 | XP_006245724.1 | 18, 19 | 4838 | |||
| XM_006245663.2 | XP_006245725.1 | 18, 19 | 5855 | |||
| XM_006245664.2 | XP_006245726.1 | 18, 19 | 3083 | |||
| XM_006245665.2 | XP_006245727.1 | 17, 18 | 4687 | |||
| XM_006245666.2 | XP_006245728.1 | 18, 19 | 2572 | |||

Lpin2
Genomic Map