siRNA Details
| Gene Symbol: Etaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109094.1 | NP_001102564.1 | 1666 | |||
| Gene Symbol: LOC316820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_229242.7 | XP_229242.2 | 1 | 1663 | ||

Etaa1
Genomic Map