siRNA Details
| Gene Symbol: Metrnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014104.1 | NP_001014126.1 | 4 | 1082 | ||
| GenBank mRNA | AB646250.1 | - | 5 | 835 | ||
| BC086590.1 | - | 4 | 1082 | 7310123 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Metrnl| Gene Symbol: Metrnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014104.1 | NP_001014126.1 | 4 | 1082 | ||
| GenBank mRNA | AB646250.1 | - | 5 | 835 | ||
| BC086590.1 | - | 4 | 1082 | 7310123 | ||