siRNA Details
| Gene Symbol: Pcdh11x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271228.1 | NP_001258157.1 | 2 | 832 | ||
| XM_008773375.1 | XP_008771597.1 | 2 | 879 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdh11x| Gene Symbol: Pcdh11x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271228.1 | NP_001258157.1 | 2 | 832 | ||
| XM_008773375.1 | XP_008771597.1 | 2 | 879 | |||