siRNA Details
| Gene Symbol: Brwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773357.1 | XP_008771579.1 | 21 | 2602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brwd3| Gene Symbol: Brwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773357.1 | XP_008771579.1 | 21 | 2602 | ||