siRNA Details
| Gene Symbol: P2ry10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177682.1 | NP_001171153.1 | 2 | 844 | ||
| XM_006257172.2 | XP_006257234.1 | 3 | 761 | |||
| XM_006257175.2 | XP_006257237.1 | 2 | 639 | |||
| XM_008773345.1 | XP_008771567.1 | 3 | 1044 | |||
| XM_008773346.1 | XP_008771568.1 | 3 | 691 | |||
| GenBank mRNA | BC168967.1 | - | 2 | 844 | 7381748 | |

P2ry10
Genomic Map