siRNA Details
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039101.1 | NP_001034190.1 | 4, 5 | 582 | ||
XM_006257145.2 | XP_006257207.1 | 4, 5 | 639 | |||
XM_008773347.1 | XP_008771569.1 | 5, 6 | 1402 | |||
XM_008773348.1 | XP_008771570.1 | 6, 7 | 555 | |||
GenBank mRNA | AY886531.1 | - | 4, 5 | 582 |
