siRNA Details
| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108245.2 | NP_001101715.1 | 5, 6 | 469 | ||
| GenBank mRNA | FM118664.1 | - | 5, 6 | 453 | ||
| FQ215476.1 | - | 5, 6 | 464 | |||
| FQ216044.1 | - | 5, 6 | 468 | |||
| FQ216100.1 | - | 5, 6 | 445 | |||
| FQ216185.1 | - | 5, 6 | 465 | |||
| FQ224925.1 | - | 5, 6 | 466 | |||

Itgb1bp2
Genomic Map