siRNA Details
| Gene Symbol: Zmym3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040155.2 | NP_001035245.2 | 7 | 1726 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001177985, NM_019831, BC139001, BC145497, AK122268, and others.

Zmym3
Genomic Map