siRNA Details
| Gene Symbol: Zmym3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040155.2 | NP_001035245.2 | 11 | 2268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmym3| Gene Symbol: Zmym3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040155.2 | NP_001035245.2 | 11 | 2268 | ||