siRNA Details
| Gene Symbol: Arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100174.1 | NP_001093644.1 | 6 | 2228 | ||
| GenBank mRNA | EF638804.1 | - | 6 | 2228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arx| Gene Symbol: Arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100174.1 | NP_001093644.1 | 6 | 2228 | ||
| GenBank mRNA | EF638804.1 | - | 6 | 2228 | ||