siRNA Details
| Gene Symbol: SLC4A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098484.2 | NP_001091954.1 | 16 | 2122 | ||
| NM_001134742.1 | NP_001128214.1 | 16 | 2122 | |||
| NM_003759.3 | NP_003750.1 | 13 | 2069 | |||
| XM_011532390.2 | XP_011530692.1 | 11 | 1608 | |||
| XM_017008792.1 | XP_016864281.1 | 14 | 1795 | |||
| XM_017008793.1 | XP_016864282.1 | 11 | 2094 | |||
| GenBank mRNA | AB470072.1 | - | 15 | 2012 | ||
| AF007216.1 | - | 13 | 2022 | |||
| AF011390.1 | - | 16 | 2122 | |||
| AF053753.1 | - | 15 | 2005 | |||
| AF053754.1 | - | 15 | 2005 | |||
| AF069510.1 | - | 15 | 2049 | |||
| AF157492.1 | - | 16 | 2081 | |||
| AF310248.1 | - | 15 | 2005 | |||
| AK226164.1 | - | 14 | 1895 | |||
| AK307594.1 | - | 16 | 2102 | |||
| EF531618.1 | - | 15 | 2005 | |||

SLC4A4
Genomic Map